Learn More
Cayman Chemical Parthenolide, 20554-84-1, 25 mg
SDP
Supplier:  Cayman Chemical 7008025
Encompass_Preferred
Molecular Formula C15H20O3, Purity ≥98%, Formula Weight 248.3; A natural sesquiterpene lactone which inhibits the growth of the promastigote form of L. amazonensis (IC50 = 3.6 g/ml); induces apoptosis in cancer cells, at least in part by inhibiting NF-B- and STAT-mediated anti-apoptotic gene transcription; directly binds the pattern recognition receptor NOD2.
Catalog No. NC1344562
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.