Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Parthenolide

Catalog No. AC462155000
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462155000 500 mg
AC462151000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC462155000 Supplier Thermo Scientific Chemicals Supplier No. 462155000

CAS: 20554-84-1 | C15H20O3 | 248.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20554-84-1
Molecular Formula C15H20O3
Molecular Weight (g/mol) 248.32
InChI Key KTEXNACQROZXEV-QLIGOWBFSA-N
IUPAC Name (1S,2S,4R,7Z,11S)-4,8-dimethyl-12-methylidene-3,14-dioxatricyclo[9.3.0.0²,⁴]tetradec-7-en-13-one
SMILES C\C1=C\CC[C@@]2(C)O[C@H]2[C@H]2OC(=O)C(=C)[C@@H]2CC1

Specifications

Melting Point 115.0°C
Color Clear
Quantity 500 mg
Merck Index 15, 7152
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in chloroform
Formula Weight 248.32
Chemical Name or Material Parthenolide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement May cause an allergic skin reaction.

Precautionary Statement GHS P Statement Wear protective gloves/protective clothing.

IF ON SKIN: Wash with plenty of soap and water.

If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.