Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Parthenolide 97.0+%, TCI America™
SDP

Catalog No. P1982100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1982100MG 100 mg
P198225MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1982100MG Supplier TCI America Supplier No. P1982100MG
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20554-84-1
Molecular Formula C15H20O3
Molecular Weight (g/mol) 248.322
MDL Number MFCD00134592
InChI Key KTEXNACQROZXEV-KIFYNCCSSA-N
Synonym --parthenolide
PubChem CID 131673949
SMILES CC1=CCCC2(C(O2)C3C(CC1)C(=C)C(=O)O3)C

Specifications

Melting Point 115°C
Color White-Yellow
Quantity 100 mg
Formula Weight 248.32
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Parthenolide
RTECSNumber LY4220000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.