Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Particle Certified HDPE Containers
GREENER_CHOICE

Catalog No. 12200117
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
1000 mL
Certified:
Low Particulate
Low Particulate, Low Metals
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Certified
12-200-117 125 mL Low Particulate
12-200-118 125 mL Low Particulate, Low Metals
12-200-140 250 mL Low Particulate
12-200-139 1000 mL Low Particulate
12-200-119 250 mL Low Particulate, Low Metals
12-200-116 1000 mL Low Particulate, Low Metals
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 12-200-117 Supplier Thermo Scientific™ Supplier No. 156125W/N/LP
Add to Cart
Add to Cart

Protect the quality of your products or lab samples with these reliable particle-certified HDPE containers,certified to contain < 20 particles per milliliter = 0.3 microns.

Protect the quality of your products or lab samples with reliable Thermo Scientific™ Particle-Certified HDPE Containers, tested and certified to contain <20 particles per milliliter = 0.3 microns. A Certificate of Analysis is included with each shipment.

A large variety of standard particle-certified containers are available. Nonstandard containers may be custom cleaned on request.

Key Features:

  • Narrow-mouth
  • Includes leak-proof† polypropylene caps
  • Suitable for use in sampling and packaging, including combination packaging for shipping of hazardous materials
  • Double-bagged and ready for cleanroom with no extra preparation

Recommended for:

Low particle applications

TRUSTED_SUSTAINABILITY

Specifications

Capacity 4 oz., 125 mL
Capacity (Metric) 125 mL
Capacity (English) 4 oz.
Color Translucent
Diameter (English) 2 in.
Material HDPE
No. per Case 24
Product Line Nalgene
Certified Low Particulate
Volume (English) Brim 4 oz.
Volume (Metric) Brim 125 mL
Certifications/Compliance Low metals certification includes aluminum, calcium, copper, iron, potassium, magnesium, manganese, sodium, and zinc at less than 10 ppb. A certificate of analysis is included with each lot.
Closure Included Yes
Mouth Narrow
Shape Round
Height (Metric) 101.6 mm
Product Type Water Sample Collection Bottle
Height (English) 4 in.
Closure Material Polypropylene
Closure Size 24-415 mm
Diameter (Metric) 50.8 mm
Workflow Step Upstream process solution storage. Upstream Validation. Upstream process solution storage
Show More Show Less

†The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.