Learn More
Cole-Parmer Environmental Devices EPAM-5000 Particulate Respiratory Health Safety Monitor
Supplier:  Cole-Parmer 1011196
Encompass
This portable, microprocessor-based particulate monitor is suitable for ambient, environmental and indoor air quality investigations. Highly sensitive, this unit scatters infrared light to measure particle concentration and provide immediate realtime determinations and data recordings of airborne particle concentration. The unique aerodynamic particle sizing and 47-mm in-line filter holder (optional accessory) loaded with the appropriate filter provide concurrent gravimetric sampling.
Catalog No. 50-630-823
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.