Description
- Compatible with CleanSpace2 PAPR Power Units
- CleanSpace™HEPA Filter comes in a pack of 3 (Pk 3)
- Used with the revolutionary CleanSpace™- A light weight PAPR with no hoses/belts
- Suitable for protection against airborne particulate (dust, mists and fumes).
- Materials: Fibreglass particulate media and plastic casing
- Easily fitted and removed from the Power Unit
Specifications
Specifications
| Product Type | Filter |
| For Use With (Equipment) | PAF-2034 |
| Quantity | 3/Pk. |