Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dentec Safety™ N95 or R95 Particulate Pad Filter Assembly
SDP

Catalog No. 19222533 Shop All Dentec Safety Specialists Inc. Products
Change view
Click to view available options
Package of 4 of each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-222-533 Package of 4 of each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-222-533 Supplier Dentec Safety™ Supplier No. 158T50
Add to Cart
Add to Cart

Particulate pads respirator filter assembly features an ultra thin design that fits under any grinding shield or welding helmet. For use with Comfort-Air™ series respirators.

  • Lightweight
  • Will not obstruct field of vision
  • Protects N and R95 filters, extending filter life
  • Saf-Twist design quickly attaches to any Dentec Safety half-mask or full face air-purifying respirator Facepiece.
  • Offset connector allows the user to rotate the cartridge into 3 different positions, improving visibility and allowing face shields or welding helmets to be worn without interference

Specifications

Color Black
Material Plastic
Product Type Filter Assembly
For Use With (Equipment) Comfort-Air™ Series
Quantity 4/Pk.
Length (Metric) 1.27 cm
Width (Metric) 8.89 cm
Height (Metric) 8.89 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.