Learn More
Description
- Contains: 5x pin hooks, 3x handle holders, 2x longish hooks
Specifications
Specifications
| Product Type | Hook Set (Standard) |
| Dimensions (L x W x H) | 14 x 11.8 x 20 in. |
| For Use With (Equipment) | VoleoPro Trolley |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.