Learn More
IBA LifeSciences pASK-IBA2C vector for E. coli with C-terminal Strep-tagII; 5 µg
Supplier:  IBA LifeSciences 21321000
Encompass_Preferred
pASK-IBA2C is a classic cloning vector devised for E. coli expression of recombinant proteins with the Strep-tagII fused to the C-terminus. It carries the chloramphenicol resistance cassette for selection of transformed E. coli cells, F1 and ColE1 origin for a high plasmid copy number, a multiple cloning site with recognition sites for several common restriction enzymes (e.g. EcoRI, SmaI, BamHI, SalI, PstI), the inducible tetracycline promoter/operator for the regulated expression of proteins, and the ompA signal for periplasmic secretion of the recombinant protein. The vector can be combined with any E. coli strain since the tet-promoter works independently of the genetic background of E. coli. Expression of the recombinant protein is induced by addition of anhydrotetracycline, which is a derivative of tetracycline that does not exhibit antibiotic activity. During secretion into the periplasmic space the ompA signal sequence will be cleaved off.
Catalog No. NC1780212
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.