Learn More
Andwin Scientific PASTEUR PIPETTES
SDP
Supplier:  Andwin Scientific 14672200
Encompass_Preferred
The disposable pasteur pipets are ideal for use in both clinical and industrial laboratories. This product is made for high-quality inoculation of culture media and transfer of samples and reagents. It has an overall length of 14.6 centimeters. This is sold in a case of 1000.
Catalog No. NC9888479
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.