Description
- Guides users through pathology imaging workflows with a digital camera and software suite
- Supports automated data generation with measurement and documentation of small biopsy tissue
- Enables standardized reporting and efficient workflow integration
- Provides real-time measurement feedback and structured gross description outputs
- Includes tools for connecting captured data to other lab systems
- Supports secure operation within facility networks for data protection compliance
- Includes a 60-day trial activation with the system and requires a valid software license for continued use
Specifications
Specifications
| Product Type | Software Subscription |
| Includes | 1 Year Service |
| For Use With (Equipment) | PathCam System |
| Control Type | Digital |