Learn More
StatLab PathFlow PiSmart Thermal Transfer Biopsy Cassettes
SDP
Catalog No. 21402253
Change view
Click to view available options
Color:
Blue
Gray
Green
Lavender
Orange
Pink
Tan
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-402-253 Blue
21-402-254 Green
21-402-255 Gray
21-402-256 Lavender
21-402-257 Orange
21-402-258 Pink
21-402-259 Tan
21-402-260 White
21-402-261 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 21-402-253 Supplier StatLab Supplier No. CASPI100BXBL
Add to Cart
Add to Cart
This item is not returnable. View return policy
PathFlow PiSmart Thermal Transfer Biopsy Cassettes, Threaded with Lid.
  • 1,000/bx, 2 bx/case, 2,000/cs
  • Threaded in strips of 40 with lids separate
  • PathFlow PiSmart Cassettes contain 4 patented pips at all 4 corners on the front of each cassette, protecting the thermal printing from high heat during dewaxing
  • Engineered specifically for use in the PiSmart Thermal Transfer Cassette printer
  • The new and improved cassettes now incorporate a smoother print face, resulting in improved print of barcodes and text
  • Available in 9 colors
Specifications
Color Blue
Product Type Biopsy Cassettes
Made in USA