Learn More
StatLab PathFlow SupaMega Slim Cassettes
Description
- SupaMega Slim cassettes uses 1/3 the depth of standard SupaMega Cassettes
- Better reagent flow and improved turnaround times
- Casting can be done in 20 minutes at -8°C
- Less wax used during the embedding process
- Large color range available
- Specific base molds
- 100 per box
- Base dimensions: 52.3 mm x 75 mm x 9.5 mm
Specifications
Specifications
| Color | Pink |
| Product Type | Large Embedding Cassette |