Learn More
Eurofins Discoverx PathHunter Anti-PL/PK Antibody, 50 μg. (92-0010)
Supplier:  Eurofins Discoverx 920010
Encompass
PathHunter Anti-ProLabel/ProLink Antibody is an antibody against the small enzyme donor (ED) fragment of β-galactosidase used in an enzyme fragment complementation assay. This antibody recognizes either the ProLabel (ProLabel) or ProLink (PK) version of ED. The antibody is a Mouse monoclonal of subclass IgG2a(κ). This antibody is provided as a vial of 50 μg of antibody in buffer at 0.5 mg/mL. For detailed product information, User manual and datasheet, visit www.discoverx.com/reagents.
Catalog No. NC0129659
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.