Learn More
Johannes Lieder Pathogenic Bacteria
SCI_ED
Catalog No. S02830 Shop All Johannes Lieder GmbH & Co. KG Products
Change view
Click to view available options
Mounting Type:
Cross and Longitudinal Section
Catalog No. Mounting Type
S02830 Cross and Longitudinal Section
1 options
Catalog No. S02830 Supplier Johannes Lieder Supplier No. 3050
Add to Cart
Add to Cart
3050 Pathogenic Bacteria, 25 microscope slides
  • 3050 Pathogenic Bacteria.
  • 25 Microscope Slides
    1. Diplococcus pneumoniae, croupous pneumonia, smear
    2. Neisseria gonorrhoeae, gonorrhoea, smear
    3. Neisseria meningitidis (intracellularis), epidemic meningitidis, smear
    4. Staphylococcus aureus, pus organism, smear
    5. Streptococcus pyogenes, smear showing short chains
    6. Corynebacterium diphtheriae, smear
    7. Mycobacterium tuberculosis, smear from positive sputum stained after Ziehl-Neelsen
    8. Bacterium erysipelatos, smear
    9. Brucella abortus, abortation in cattle (Bang disease), smear
    10. Proteus vulgaris, inflammation of urinary system, smear
    11. Escherichia coli, colon bacteria, possibly pathogen, smear
    12. Eberthella typhi, typhoid fever, smear
    13. Salmonella paratyphi, paratyphoid fever, smear
    14. Hemophilus influenzae (Pfeiffer), smear
    15. Klebsiella pneumoniae (Friedlander), pneumonia, smear
    16. Pasteurella (Yersinia) pestis, bubonic plague, smear
    17. Salmonella enteritidis, meat poisoning, smear
    18. Shigella dysenteriae, bacillary dysentery, smear
    19. Bacillus anthracis, wool sorter's disease, smear
    20. Clostridium botulinum, food poisoning, smear
    21. Clostridium septicum, smear
    22. Clostridium tetani, lockjaw, smear
    23. Clostridium perfringens, gas gangrene, smear
    24. Vibrio comma, Asiatic cholera, smear
    25. Borrelia duttoni (Spirochaeta recurrentis), Central African relapsing fever, blood smear
Specifications
Classification Microbiology
Material Elastin Stained Glass
Mounting Type Cross and Longitudinal Section
Type Microscope Slides
Slide Type Prepared Slide