Learn More
Laxco™ Pathology Upgrade Kit for SeBaP4C2
Pathology upgrade kit for analyzing various pathological sample slides and diagnosis.
Supplier: Laxco™ SEBAP4C2PATKIT
Description
- 1.25x APO objective,1.2 NA swing out condenser, plan achromat 60x objective.
- Ideal for observing a larger are of the slide in a single view.
- Suitable for tissue samples extracted during biopsy or surgery.
Specifications
Adapter Kit | |
SEBAP4C2 Microscope | |
100/115/220/230 V |
Not Autoclavable | |
Sterile |