Learn More
BD Patient/Exam Room Sharps Collectors
Catalog No. 14826142 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Capacity (English):
5.4 qt.
2 gal.
3 gal.
Color:
Clear
Pearl
Red
Design:
Counterbalanced Door
Counterbalanced Side Entry
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Color Design
14-826-142 2 gal. Pearl Counterbalanced Door
14-826-145 3 gal. Red Counterbalanced Door
14-826-65A 5.4 qt. Red Counterbalanced Door
14-830-29 5.4 qt. Pearl Counterbalanced Side Entry
14-826-64B 5.4 qt. Clear Counterbalanced Side Entry
14-826-65B 5.4 qt. Clear Counterbalanced Door
14-826-144 2 gal. Red Counterbalanced Door
14-830-28 5.4 qt. Red Counterbalanced Side Entry
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 14-826-142 Supplier BD Supplier No. 305422
Add to Cart
Edge
Add to Cart
Horizontal, side or vertical entry
  • Strong, durable polypropylene construction gives containers excellent impact and puncture-resistance
  • Suitable for incineration or autoclaving
  • Clear top and door provide visual indication of fill level
  • Safety transport handles keep hands away from collector body
Horizontal- and Side-Entry Collectors
  • Counterbalanced door opens automatically for single-handed disposal; stays closed to prevent overfilling
Order Info
Shipped nested; assembly of top to base required.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 7.5 L
Closure Color Pearl
Design Counterbalanced Door
Capacity (English) 2 gal.
Color Pearl
Vent No
Material Polypropylene