Learn More
Description
- Strong, durable polypropylene construction gives containers excellent impact and puncture-resistance
- Suitable for incineration or autoclaving
- Clear top and door provide visual indication of fill level
- Safety transport handles keep hands away from collector body
Horizontal- and Side-Entry Collectors
- Counterbalanced door opens automatically for single-handed disposal; stays closed to prevent overfilling
Order Info
Shipped nested; assembly of top to base required.
Specifications
Specifications
| Capacity (Metric) | 5.7 L |
| Design | Counterbalanced Side Entry |
| Capacity (English) | 5.4 qt. |
| Color | Clear |
| Disposable | Single-use |
| For Use With (Application) | Suitable for incineration or autoclaving |
| Material | Polypropylene |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.