Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Patulin, 99%

Catalog No. p-178335
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC228870050 5 mg Glass bottle
AC228870100 10 mg Glass bottle
AC228870250 25 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC228870050 Supplier Thermo Scientific Chemicals Supplier No. 228870050
Only null left
Add to Cart

CAS: 149-29-1 | C7H6O4 | 154.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-29-1
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.12
MDL Number MFCD00005858
InChI Key ZRWPUFFVAOMMNM-UHFFFAOYSA-N
Synonym patulin, clavacin, expansin, clavatin, claviform, expansine, patuline, clairformin, claviformin, gigantin
PubChem CID 4696
ChEBI CHEBI:74926
IUPAC Name 4-hydroxy-4,6-dihydrofuro[3,2-c]pyran-2-one
SMILES C1C=C2C(=CC(=O)O2)C(O1)O

Specifications

Melting Point 108.0°C to 111.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 mg
Merck Index 15, 7158
Solubility Information Solubility in water: soluble. Other solubilities: soluble in most common organic solvents,soluble in ethyl and amyl acetate,insoluble in petroleum ether
Formula Weight 154.12
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Patulin, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-735-2
RTECSNumber LV2625000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.