Learn More
Paulson A-TAC™ Firefighter Goggles: Structural
SDP
Catalog No. 19816409 Shop All Paulson Mfg Products
Change view
Click to view available options
Product Type:
Firefighter Goggles
Firefighter goggle w/quick relaese buckle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-816-409 Firefighter goggle w/quick relaese buckle
19-816-412 Firefighter Goggles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-816-409 Supplier Paulson Supplier No. 510EB
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed for protection when fighting wild or structural fires.
  • Polycarbonate, hard-coated/anti-fog lens
  • Fit comfortably over prescription, sun, and safety glasses
  • Indirect ventilation ports
  • Can be placed on the helmet or worn on the head
Model 510EB
  • Silicone frame
  • Optically correct lens
  • Quick-release buckle
  • Apec lens
  • Black frame
Model 510SL
With noseguard
Compliance
Meets ANSI Z87.1 standard for impact, penetration and chemical splash exposure
Specifications
Certifications/Compliance ANSI Z87.1
Description Apec lens
Lens Tint Clear
Product Type Firefighter goggle w/quick relaese buckle
For Use With (Application) Wildland, structural or intense heat fire fighter
Frame Color Black