Learn More
Paulson Corrections Faceshield
SDP
Catalog No. 19500359 Shop All Paulson Mfg Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
19-500-359 Clear
1 options
Catalog No. 19-500-359 Supplier Paulson Supplier No. DK5H150/CFS
Fit standard military-style and most police helmets
Helmet-mounted faceshield with Lexan™ window and top rubber seal to prevent seapage of liquids onto wearer's face. Faceshield has 0.150 in.-thick window and adds spring-steel-wire grid for increased protection against aggresive attacks.
  • Pivot-and-lock mechanism mounts to either side of shield allowing left- or right-handed operation
  • Shield locks up or down and stays in place
  • Extra clearance for use over gas mask
  • Field-mounted style with mounting straps to adapt to various types of helmets
Compliance
Meets NIJ0104.02.
Specifications
Product Type Field Mounted Correction Faceshield
Color Clear