Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paulson Hat Bracket for Face Shield
SDP

Catalog No. 19816423 Shop All Paulson Mfg Products
Change view
Click to view available options
For Use With (Equipment):
Universal 7-pattern Slot Hole Window
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-816-423 Universal 7-pattern Slot Hole Window
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-816-423 Supplier Paulson Supplier No. 9102401

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Compatible with Paulson Industrial Face Shield (19-816-430).

Specifications

Color Blue
For Use With (Equipment) Universal 7-pattern Slot Hole Window
For Use With (Application) Dielectric and high temperature utility
Includes Five cam locks, Adjustable pivots
Material Nylon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.