Learn More
Paulson Industrial Faceshield
SDP
Catalog No. 19822651 Shop All Paulson Mfg Products
Change view
Click to view available options
Industrial faceshield, polycarbonate, 9.25 x 15.5 x 0.040 in. (23.4 x 39.3 x 0.1cm)
Catalog No.
19-822-651 Industrial faceshield, polycarbonate, 9.25 x 15.5 x 0.040 in. (23.4 x 39.3 x 0.1cm)
1 options
This item is not returnable. View return policy
Catalog No. 19-822-651 Supplier Paulson Supplier No. S25L4
This item is not returnable. View return policy
Individual inspection provides consistent quality and high optical standards
  • Flat lexan industrial face shield
  • Attachment pattern consists of center hole with 4 slots
  • Clear window made of polycarbonate
  • 4-holes slot from center designed to fit PAULSON head gear and helmet brackets
  • Designed to meet ANSI Z87.1 Low Impact
  • Dimensions: 9.25 x 15.5 x 0.040 in. (23.4 x 39.3 x 0.1cm) in.
Specifications
Lens Tint Clear