Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paulson KneeShield
SDP

Catalog No. 19500334 Shop All Paulson Mfg Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-500-334 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-500-334 Supplier Paulson Supplier No. 6258340

missing translation for 'validMainframeMsg'

For use in corrections-inmate confrontations and control

Rigid shell knee pad with heavy elastic and hook-and-loop adjustment. Protects knee from above and below beyond standard style knee pads.

  • Articulated for comfortable movement
  • Resists penetration from sharp or pointed objects and impact
  • Designed to be worn loosely for comfort and quickly tightened before conflict

Order Info

Weight: 16 oz. Black.

Specifications

Color Black
Material Elastic
For Use With (Application) Corrections-inmate confrontations and control
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.