Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paulson Lexan™ Standard Body Shield
SDP

Catalog No. 19500343 Shop All Paulson Mfg Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-500-343 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-500-343 Supplier Paulson Supplier No. BS2

missing translation for 'validMainframeMsg'

Unique hardware mounting system reduces likelihood of shield cracking

Constructed of clear, impact-resistant Lexan, body shields feature ergonomic handle allowing for two handed use in extreme circumstances.

  • Dielectric handle and break away strap prevent electrical pass-through
  • Handle can be reversed for right- or left-handed use
  • Wide nylon arm strap adjusts easily while in use
  • Superior optical quality and workmanship

Order Info

Shields shipped unassembled. Available with choice of ID label: Police (available in English or Spanish), State Police, Federal Police, Military Police, International Military Police, Sheriff, Security, and Corrections, which must be specified at time of order.

Specifications

Product Type Body Shield
Color Clear
Material Polycarbonate, Polymer Alloy with Steel Inserts
For Use With (Application) Police, sheriff, national guard, university police, riot squad, jail
Height (English) 36 in.
Includes ID label
Size Standard
Weight (English) 6 lb.
Width (English) 20 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.