Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paulson Shield Labels
SDP

Catalog No. 19500355 Shop All Paulson Mfg Products
Change view
Click to view available options
Model:
Corrections Label
Military Police Label
Police Label
Policia Label
Sheriff Label
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
19-500-355 Sheriff Label
19-500-351 Corrections Label
19-500-353 Police Label
19-500-352 Military Police Label
19-500-354 Policia Label
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-500-355 Supplier Paulson Supplier No. BSLABS

missing translation for 'validMainframeMsg'

Provide positive identification at a distance

  • Replacement labels for body and capture shields
  • Mylar™ self-adhesive labels attach to inside surface of shield

Specifications

Product Type Body Shield Identification Label
Color Black on White
Material Mylar
Diameter (Metric) 60.96 cm
For Use With (Equipment) Body and capture shield
Model Sheriff Label
Weight (English) 4 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.