Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paulson Sun Shield for MSA™ Top-Guard Hat
SDP

Catalog No. 19816418 Shop All Paulson Mfg Products
Change view
Click to view available options
Transparent front section; Color: White/Smoke
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-816-418 Transparent front section; Color: White/Smoke
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-816-418 Supplier Paulson Supplier No. 1615740

missing translation for 'validMainframeMsg'

Specifications

Color Smoke, White
For Use With (Equipment) MSA Top-Guard Hat
For Use With (Application) UV protection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.