Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paulson Sun Shield for MSA™ V-Guard Hat
SDP

Catalog No. 19816406 Shop All Paulson Mfg Products
Change view
Click to view available options
For Use With (Equipment):
MSA V-Gard Cap-style Hard Helmet
MSA V-Gard Full Brim Hard Hats
Product Type:
Full Brim Hard Hat Sun Shield
Sun Shield
For Use With (Application):
Nose and neck protection against sun light for people wearing a helmet at outdoor activities
Includes:
Transparent front part and UV protective sun glare filter, clamps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Product Type For Use With (Application) Includes
19-816-406 MSA V-Gard Cap-style Hard Helmet Sun Shield Nose and neck protection against sun light for people wearing a helmet at outdoor activities Transparent front part and UV protective sun glare filter, clamps
19-816-416 MSA V-Gard Full Brim Hard Hats Full Brim Hard Hat Sun Shield - -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-816-406 Supplier Paulson Supplier No. 1615640
Add to Cart
Add to Cart

Specifications

Certifications/Compliance DIN EN 166:2002, DIN EN 172Annex II of the PPE-Directive89/686/EEC, ANSI Z87.1
Color Smoke, White
For Use With (Equipment) MSA V-Gard Cap-style Hard Helmet
For Use With (Application) Nose and neck protection against sun light for people wearing a helmet at outdoor activities
Includes Transparent front part and UV protective sun glare filter, clamps
Length (Metric) Face Shield 6 cm
Length (English) Face Shield 2.4 in.
Material ABS, Lexan
Product Type Sun Shield
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.