Learn More
Biocare Medical, LLC PAX-5 Antibody - Concentrate, 0.5 ml
SDP
Supplier:  Biocare Medical, LLC CM207B
Encompass_Preferred
PAX-5 Antibody is a B-cell specific activator protein (BSAP). In early stages of B-cell development, PAX-5 influences the expression of several B-cell specific genes, such as CD19 and CD20. PAX-5 is expressed primarily in pro-, pre-, and mature B-cells, but not in plasma cells. There is an excellent correlation between CD20 and Pax-5 expression; however anti-Pax-5 exceeds the specificity and sensitivity of L26 (CD20) because of its earlier expression in B-cell differentiation and its ability to detect all committed B-cells, including classic Hodgkin lymphoma. It is very specific to B-cell lineage and does not stain T-cells. In essence, PAX-5 may be a superior pan B-cell marker to CD20 (see reference).
Catalog No. 50-826-00
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.