Learn More
Invitrogen™ PAX1 Polyclonal Antibody

Description
PAX1 Polyclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | PAX1 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | PAX1 |
| Gene Accession No. | P15863 |
| Gene Alias | hbs; hunchback; HuP48; OFC2; paired box 1; paired box gene 1; Paired box homeotic gene 1; paired box protein Pax-1; paired domain gene HuP48; PAX1; Pax-1; un; undulated; wavy tail; wt |
| Gene Symbols | PAX1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.