Learn More
PAX8 Antibody, Novus Biologicals™

Description
Specifications
Specifications
| Antigen | PAX8 |
| Applications | Chemotaxis, ChIP Assay, ChIP Assay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Simple Western, Single Cell Western, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:5000-1:20000, Simple Western 1:10, Immunohistochemistry 1:100 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunoprecipitation 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:100 - 1:1000, Chromatin Immunoprecipitation (ChIP), Single Cell Western 50 ug/mL |
| Formulation | 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal |
| Gene Alias | paired box 8, paired box gene 8, paired box protein Pax-8, paired domain gene 8 |
| Gene Symbols | PAX8 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.