Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAX9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572650
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572650 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572650 Supplier Invitrogen™ Supplier No. PA572650
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PAX9 Polyclonal Antibody for Western Blot

This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The specific function of the paired box 9 gene is unknown but it may involve development of stratified squamous epithelia as well as various organs and skeletal elements.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAX9
Applications Western Blot
Classification Polyclonal
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene pax9
Gene Accession No. 0
Gene Alias EMBL:BAE79272.1}; etID309776.17; paired box 9; paired box gene 9; paired box homeotic gene 9; paired box protein Pax-9; paired domain gene 9; Pax9; Pax-9; pax9 {ECO:0000312; pax9 protein; Pax9a; Pax9b; STHAG3
Gene Symbols pax9
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 69-95 amino acids from the N-terminal region of Zebrafish Pax9.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30558
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.