Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAX9 Recombinant Rabbit Monoclonal Antibody (SD082-02)
GREENER_CHOICE

Catalog No. PIMA532380
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA532380 100 μL
1 options

Catalog No. PIMA532380

Supplier: Invitrogen™ MA532380

Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The specific function of the paired box 9 gene is unknown but it may involve development of stratified squamous epithelia as well as various organs and skeletal elements.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAX9
Applications Western Blot
Classification Recombinant Monoclonal
Clone SD082-02
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene pax9
Gene Accession No. P47242, P55771, Q2L4T2
Gene Alias EMBL:BAE79272.1}; etID309776.17; paired box 9; paired box gene 9; paired box homeotic gene 9; paired box protein Pax-9; paired domain gene 9; Pax9; Pax-9; pax9 {ECO:0000312; pax9 protein; Pax9a; Pax9b; STHAG3
Gene Symbols pax9
Host Species Rabbit
Immunogen Recombinant protein within Human PAX9 aa 6-120.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18511, 362741, 5083
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.