Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Paxillin Monoclonal Antibody (PXC-10)
GREENER_CHOICE

Catalog No. PIMA124952 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA124952 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA124952 Supplier Invitrogen™ Supplier No. MA124952
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

MA1-24952 detects Paxillin from human, rat, bovine, chicken, and hamster samples.

Paxillin is a focal adhesion protein and a substrate for several tyrosine kinases such as src, FAK, and p210BRC/ABL. The tyrosine phosphorylation of paxillin is affected by conditions that change cell-cell adhesion. Paxillin associates tightly with FAK and Crk through its SH2 domain. This interaction is independent of the extracellular matrix. Although paxillin was initially discovered in fibroblasts, its phosphorylation may also be important during neurite extension during differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Paxillin
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone PXC-10
Concentration 5.9 mg/mL
Conjugate Unconjugated
Formulation Ascites with 15mM sodium azide
Gene PXN
Gene Accession No. P49023, P49024, Q66H76, Q8VI36
Gene Alias adaptor protein; AW108311; AW123232; FLJ16691; hCG_1778014; I79_001229; myocardial ischemic preconditioning associated protein 7; Pax; PAX-1; PAXI; Paxillin; paxillin a; paxillin alpha; pxn; pxna; testicular tissue protein Li 134; TGFB1I1; transforming growth factor beta 1 induced transcript 1; wu:fw71f12
Gene Symbols PXN
Host Species Mouse
Immunogen C-terminal part of recombinant chicken paxillin (amino acids 305-559).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100760740, 19303, 360820, 395832, 517456, 5829
Target Species Human, Mouse, Rat, Bovine, Hamster, Chicken
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.