Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Paxilline, Ca2+ Activated K+ Channel Blocker, Solid, 1 mg

Catalog No. 502706416
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6416 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6416 Supplier Abcam Supplier No. AB1418401MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent, high-conductance Ca2+-activated K+channel blocker with anticonvulsant effects in vivo.

  • Potency: Potent high-conductance Ca2+-activated K+ channel blocker (Ki = 2 nM, α-subunit BKCa)
  • Inhibition: Inhibits sarco/endoplasmic reticulum Ca2+-ATPase (IC50 = 5 to 50 μM)
  • Purity: >98%
  • Molecular Weight: 435.6 Da
  • Biological Activity: Shows anticonvulsant effects in vivo
  • Form: Solid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Target Molecule Ca2+-activated K+ Channel, Sarco/endoplasmic Reticulum Ca2+-ATPase
Product Type Blocker and Diluent
Buffer 100% Paxilline
Content And Storage Store at -20°C.
Form Solid
Formulation 100% Paxilline
Quantity 1 mg
For Use With (Application) Ca2+-activated K+channel blocking, sarco/endoplasmic reticulum Ca2+-ATPase inhibition, anticonvulsant research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.