Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Paxilline, Ca2+ activated K+ Channel Blocker, Solid, 5 mg
Description
- Molecular Weight: 435.6 Da
- Purity: >98%
- Physical State: Solid
- Activity: Potent high-conductance Ca2+-activated K+ channel blocker (Ki = 2 nM, α-subunit BKCa)
- Secondary Activity: Inhibits sarco/endoplasmic reticulum Ca2+-ATPase (IC50 = 5 to 50 μM)
- Biological Effect: Shows anticonvulsant effects in vivo
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Product Type | Blocker and Diluent |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Shipping Condition | Ambient |
| Target Molecule | Ca2+-activated K+ Channels, Sarco/Endoplasmic Reticulum Ca2+-ATPase |
| Formulation | 100% Paxilline |
| Quantity | 5 mg |
| For Use With (Application) | Ca2+-activated K+channel blocking, SERCA inhibition, anticonvulsant research |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?