Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAXIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552327
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552327 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552327 Supplier Invitrogen™ Supplier No. PA552327
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EEEEEVENEE QDSQNEGSTD EKSSPASSQE GSPSGDQQFS PKSNTEKSKG ELMFDDSSDS SPEKQERNLN WTPAEVPQLA AAKRRLPQGK EPGLINLCAN VPPVPGNILP PEVRGNLMAA GQNLQSSERS Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 50%.

This gene is a member of the paired box gene family and encodes a nuclear protein with six BRCT domains. This protein plays a critical role in maintaining genome stability, condensation of chromatin and progression through mitosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAXIP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PAXIP1
Gene Accession No. Q6ZW49
Gene Alias CAGF28; CAGF29; D5Ertd149e; fa04b06; FLJ41049; PACIP1; PAX interacting (with transcription-activation domain) protein 1; PAX interacting protein 1; PAX transactivation activation domain-interacting protein; PAX transcription activation domain interacting protein 1 like; PAX-interacting protein 1; PAX-interacting protein 1-like protein; Paxip1; PAXIP1L; protein encoded by CAG trinucleotide repeats; PTIP; TNRC2; wu:fa04b06; zgc:92593
Gene Symbols PAXIP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PAXIP1. Recombinant protein control fragment (Product #RP-89488).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22976
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.