Learn More
TARGETMOL CHEMICALS INC PAZOPANIB 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T0097L
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pazopanib (GW786034) a small molecule inhibitor inhibits multiple protein tyrosine kinases with potential antineoplastic activity. Pazopanib selectively inhibits VEGFR-1 -2 and -3 c-kit and PDGF-R which may result in inhibition of angiogenesis in tumors in which these receptors are upregulated. purity: 99%
Catalog No. 50-258-1796
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.