Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pazufloxacin Mesylate 98.0+%, TCI America™
SDP

Catalog No. p-7135334 Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19621G 1 g
P19625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P19621G Supplier TCI America Supplier No. P19621G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 163680-77-1
Molecular Formula C17H19FN2O7S
Molecular Weight (g/mol) 414.404
MDL Number MFCD00913262
InChI Key UDHGFPATQWQARM-FJXQXJEOSA-N
Synonym Pazufloxacin Methanesulfonate
PubChem CID 6918232
SMILES CC1COC2=C3N1C=C(C(=O)C3=CC(=C2C4(CC4)N)F)C(=O)O.CS(=O)(=O)O

Specifications

Melting Point 259°C
Color Yellow
Quantity 1 g
Formula Weight 414.40
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pazufloxacin Mesylate
RTECSNumber UU8815400
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.