Learn More
Biolegend Pacific Blue™ anti-human CD73 (Ecto-5'-nucleotidase)
Supplier:  Biolegend 344012
Encompass_Preferred
Pacific Blue™ anti-human CD73 (Ecto-5'-nucleotidase) [AD2]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon; Apps: FC; Size: 100 tests
Catalog No. 50-207-6608