Learn More
Charles River Labs Cell Solutions Peripheral Blood Gamma/Delta T Cells, 2 Million cells, Cryopreserved
Supplier:  Charles River Labs Cell Solutions PB03GDC1
Encompass_Preferred
Gamma delta T cells are in low abundance in the body and are found in the gut mucosa, skin, lungs, and uterus. These cells are involved in the initiation and propagation of immune responses. They are considered to represent a link between innate and adaptive immunity. They can secrete particular effector cytokines in a subtype- and context-specific manner and are capable of phagocytosis. CD3+ T-cell receptor γ/δ+ (TCRγ/δ+) cells are negatively selected via immunomagnetic bead separation from a leukapheresis collection.
Catalog No. 50-225-6288
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.