Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PBDC1 Monoclonal Antibody (OTI5A11)
GREENER_CHOICE

Catalog No. PIMA525600
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525600 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525600 Supplier Invitrogen™ Supplier No. MA525600
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PBDC1 Monoclonal Antibody for Western Blot, IHC (P)

PBDC1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PBDC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5A11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Pbdc1
Gene Accession No. Q9BVG4
Gene Alias 2610029G23Rik; CXorf26; MGC874; Pbdc1; polysaccharide biosynthesis domain containing 1; polysaccharide biosynthesis domain-containing protein 1; Protein PBDC1; UPF0368 protein Cxorf26; UPF0368 protein Cxorf26 homolog
Gene Symbols Pbdc1
Host Species Mouse
Immunogen Full length human recombinant protein of CXorf26 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51260
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.