Learn More
PBDEs in H2O, PT Proficiency Testing Material, MilliporeSigma™ Supelco™
Catalog No. 111017328 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 mL
Catalog No. Quantity
11-101-7328 2 mL
1 options
Catalog No. 11-101-7328 Supplier MilliporeSigma Supelco Supplier No. PE13982ML
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sampleThis PT is valid for ISO method 22032.
This PT sample is packaged as a 1.5 mL concentrate. The PT sample is not preserved. The sample must be stored at 4 CSample preparationAllow the ampule concentrate to equilibrate to room temperature. Fill a 1000 mL Class A Volumetric Flask with 900 mL of organic free reagent water. Open the ampule and pipet exactly 1.0 mL of the concentrate into the flask. Fill to volume and mix well. This represents the sample for analysis.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Ampule
Quantity 2 mL
Concentration or Composition (by Analyte or Components) BDE 153 20-500 ng/L; BDE 99 20-500 ng/L; BDE 100 20-500 ng/L; BDE 47 20-500 ng/L; BDE 154 20-500 ng/L; BDE 183 20-500 ng/L; BDE 205 20-500 ng/L; BDE 209 20-500 ng/L