Learn More
Abcam PBFI AM, Fluorescent K+ Binding Probe, Solid, 1 mg
Description
- Molecular Weight: 1171.1 Da
- Purity: >94%
- Form: Solid
- Properties: Cell-permeable, fluorescent K+ binding probe (Kd = 5 mM)
- Activity: Active in vitro
- Mechanism: Non-specific esterases hydrolyze AM esters to produce the water-soluble form of PBFI
- Excitation: 340/390 nm
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Target Molecule | Potassium (K+) |
| Label or Dye | Fluorescent Probe |
| Form | Solid |
| Detection Method | Fluorescence |
| Content And Storage | Store at -20°C. |
| Detection Location | In-Cell Detection, In-Solution Detection |
| Formulation | 100% PBFI AM |
| Quantity | 1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.