Learn More
Promega pBiT3.1-N[CMV/HiBiT/Blast] Vector, 20microg
Catalog No. PRN2361 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
pBiT3.1-N[CMV/HiBiT/Blast] Vector, 20microg
Catalog No.
PRN2361 pBiT3.1-N[CMV/HiBiT/Blast] Vector, 20microg
1 options
This item is not returnable. View return policy
Catalog No. PRN2361 Supplier Promega Supplier No. N2361
This item is not returnable. View return policy
The pBiT3.1-N, pBiT3.1-C and pBiT3.1-secN [CMV/HiBiT/Blast] Vectors are traditional cloning vectors for inserting proteins of interest. Cloned ORFs contain the HiBiT tag at the N or C terminus; the N terminus for transmembrane or secreted proteins.
  • Traditional Cloning Vectors for Generating HiBiT−Tagged Proteins
  • Add an N− or C−terminal HiBiT tag to your protein of interest
  • Generate an N−terminal HiBiT−tagged transmembrane or secreted protein
  • Use for expression analysis and protein quantitation using luminescent detection reagents