Learn More
Promega pBiT3.1-secN[CMV/HiBiT/Blast] Vector, 20microg
Catalog No. PRN2381 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
20 μg
Catalog No. Quantity
PRN2381 20 μg
1 options
This item is not returnable. View return policy
Catalog No. PRN2381 Supplier Promega Supplier No. N2381
This item is not returnable. View return policy
The pBiT3.1-N, pBiT3.1-C and pBiT3.1-secN [CMV/HiBiT/Blast] Vectors are traditional cloning vectors for inserting proteins of interest. Cloned ORFs contain the HiBiT tag at the N or C terminus; the N terminus for transmembrane or secreted proteins.
  • Traditional Cloning Vectors for Generating HiBiT−Tagged Proteins
  • Add an N− or C−terminal HiBiT tag to your protein of interest
  • Generate an N−terminal HiBiT−tagged transmembrane or secreted protein
  • Use for expression analysis and protein quantitation using luminescent detection reagents
Specifications
Content And Storage -30°C to -10°C
Quantity 20 μg