Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Glutamic Acid, FCC, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18603834 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
12 kg
25 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-834 12 kg Poly Bottle
18-603-830 100 g Poly Bottle
18-603-832 25 kg Poly Bottle
18-603-833 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-603-834 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. G103612KG
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

G1036, 56-86-0, C5H9NO4

Spectrum™ Chemical L-Glutamic Acid, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 56-86-0
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00002634
InChI Key WHUUTDBJXJRKMK-UHFFFAOYNA-N
IUPAC Name 2-aminopentanedioic acid
SMILES NC(CCC(O)=O)C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.3%
Loss on Drying 0.1%
Packaging Poly Bottle
Quantity 12 kg
Optical Rotation +31.5° to +32.5°
Percent Purity 98.5 to 101.5%
Grade FCC
Physical Form Crystalline Powder or Lumps
Chemical Name or Material L-Glutamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.