Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PBK Recombinant Rabbit Monoclonal Antibody (K01_1W40)

Catalog No. MA564864
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA564864 50 μL
MA563405 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA564864 Supplier Invitrogen™ Supplier No. MA564864
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: PSAAHIVEAL ETDV.

PBK (TOPK) is a serine/threonine kinase that is related to the dual specific mitogen-activated protein kinase (MAPKK). The kinase is up-regulated in a variety of hematological malignancies, participates in activating lymphoid cells, and supports testicular function (spermatogenesis). Evidence suggests that mitotic phosphorylation is required for its catalytic activity. This mitotic kinase may be involved in the activation of lymphoid cells and support testicular functions, with a suggested role in the process of spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PBK
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_1W40
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene PBK
Gene Accession No. Q96KB5, Q9JJ78
Gene Alias 2810434B10Rik; AW538537; Cancer/testis antigen 84; CT84; D14Ertd732e; epididymis luminal protein 164; HEL164; Lymphokine-activated killer T-cell-originated protein kinase; MAPKK-like protein kinase; Nori-3; PBK; PDZ binding kinase; PDZ-binding kinase; serine/threonine protein kinase; spermatogenesis-related protein kinase; SPK; T-LAK cell-originated protein kinase; TOPK
Gene Symbols PBK
Host Species Rabbit
Immunogen A synthetic peptide of human PBK (aa 309-322).
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 52033, 55872
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.