Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PBRM1 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993358
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3358 100 μL
50-199-3336 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3358 Supplier Active Motif Supplier No. 61381
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Involved in transcriptional activation and repression of select genes by chromatin remodeling (alteration of DNA-nucleosome topology).

Specifications

Antigen PBRM1
Applications ChIP sequencing (ChIP-seq), Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene Pbrm1
Gene Accession No. Q86U86, Q8BSQ9
Gene Alias 2310032M22Rik; 2610016F04Rik; AI507524; BAF180; BOS_21051; BRG1-associated factor 180; hPB1; PB1; PBRM1; polybromo 1; Polybromo-1D; Protein polybromo-1; RGD1565549
Gene Symbols Pbrm1
Host Species Rabbit
Immunogen This antibody was raised against a peptide within the N-terminal region of human PBRM1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55193, 66923
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.