Learn More
Cytiva PBS Buffer 10, 1000mL
Catalog No. 501055347 Shop All Cytiva Products
Encompass
Contents: 0.1 M phosphate buffer with 27 mM KCl and 1.37 M NaCl, 1 1000 ml; Running buffer. Concentrated stock solution, yielding pH 7.4 when diluted 10X and supplemented with 5% DMSO. (PBS buffer with DMSO is commonly used for small molecule assays.)
Catalog No. 50-105-5347 Supplier Cytiva Supplier No. BR100672
Contents: 0.1 M phosphate buffer with 27 mM KCl and 1.37 M NaCl, 1 1000 ml; Running buffer. Concentrated stock solution, yielding pH 7.4 when diluted 10X and supplemented with 5% DMSO. (PBS buffer with DMSO is commonly used for small molecule assays.)